(R)-2-(Pyrrolidin-3-yl)propan-2-ol hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A1469658-100MG
A1469658-1G
A1469658-250MG
A1469658-5G
76662-676EA
44
USD
76662-676
76662-678
76662-680
77622-208
(R)-2-(Pyrrolidin-3-yl)propan-2-ol hydrochloride 97%
(R)-2-(Pyrrolidin-3-yl)propan-2-ol hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₆ClNO |
MDL Number:
MFCD31801468 CAS Number: 2203403-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...