4-(But-3-yn-1-ylamino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 97%

Supplier: Ambeed
Warning

A1469637-1G A1469637-250MG
76662-672EA 211 USD
76662-672 76662-674
4-(But-3-yn-1-ylamino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 97%
4-(But-3-yn-1-ylamino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₇H₁₅N₃O₄
CAS Number: 2154356-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR