BMOE (bis-Maleimidoethane) 98%

Supplier: Ambeed
Warning

Synonyms: Ethylene-bis-maleimide, 1,2-Bis-maleimidoethane, 1,2-Bismaleimidoethane, 1,2-Di(maleimido)ethane, N,N'-Ethylenebismaleimide, 1,2-Bis(maleimido)ethane, 1,1'-(Ethane-1,2-diyl)bis(1H-pyrrole-2,5-dione), BMOE, N,N'-Ethylenedimaleimide

A1468003-100MG A1468003-1G A1468003-250MG
76662-660EA 30.77 USD
76662-660 76662-662 76662-664
BMOE (bis-Maleimidoethane) 98%
BMOE (bis-Maleimidoethane)

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₄
MW: 220.18 g/mol
MDL Number: MFCD00458699
CAS Number: 5132-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR