3-Methoxy-4-(methoxycarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Danger

A1467993-100MG A1467993-1G A1467993-250MG A1467993-25G A1467993-5G
76662-644EA 6 USD
76662-644 76662-646 76662-648 76662-650 76662-652
3-Methoxy-4-(methoxycarbonyl)phenylboronic acid 98%
3-Methoxy-4-(methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₅
MDL Number: MFCD07366483
CAS Number: 603122-41-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR