Dimethyl 4,4'-([2,2'-bipyridine]-5,5'-diyl)dibenzoate 97%
Supplier: Ambeed
|
Warning
|
A1467897-100MG
A1467897-1G
A1467897-250MG
76662-618EA
92
USD
76662-618
76662-620
76662-622
Dimethyl 4,4'-([2,2'-bipyridine]-5,5'-diyl)dibenzoate 97%
Dimethyl 4,4'-([2,2'-bipyridine]-5,5'-diyl)dibenzoate
Precaución: for Research Use Only
Formula:
C₂₆H₂₀N₂O₄ |
CAS Number:
1373759-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...