2-Bromo-4-chloronicotinaldehyde 97+%

Supplier: Ambeed
Danger

A1467848-100MG A1467848-1G A1467848-250MG A1467848-25G A1467848-5G
76662-608EA 9 USD
76662-608 76662-610 76662-612 76662-614 76662-616
2-Bromo-4-chloronicotinaldehyde 97+%
2-Bromo-4-chloronicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃BrClNO
MDL Number: MFCD20040081
CAS Number: 1289197-78-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR