2,6-Dibromonicotinaldehyde 97%

Supplier: Ambeed
Warning

A1467836-1G A1467836-250MG A1467836-5G
76662-602EA 44 USD
76662-602 76662-604 76662-606
2,6-Dibromonicotinaldehyde 97%
2,6-Dibromonicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃Br₂NO
MDL Number: MFCD10574710
CAS Number: 55304-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR