1,2-Bis(4-nitrophenyl)ethyne 98%

Supplier: Ambeed
Warning

A1467624-1G A1467624-250MG
76662-580EA 163.38 USD
76662-580 76662-582
1,2-Bis(4-nitrophenyl)ethyne 98%
1,2-Bis(4-nitrophenyl)ethyne

Precaución: for Research Use Only

Formula: C₁₄H₈N₂O₄
MW: 268.2243 g/mol
MDL Number: MFCD00605326
CAS Number: 2735-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR