6,7-Dimethoxyquinazoline-2,4-diamine dihydrochloride 95%

Supplier: Ambeed
Warning

A1467338-100MG A1467338-1G A1467338-250MG
76662-548EA 45 USD
76662-548 76662-550 76662-552
6,7-Dimethoxyquinazoline-2,4-diamine dihydrochloride 95%
6,7-Dimethoxyquinazoline-2,4-diamine dihydrochloride
Formula: C₁₀H₁₄Cl₂N₄O₂
MDL Number: MFCD34186962
CAS Number: 2703752-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR