2,4,7-Trichloro-8-fluoropyrido[4,3-d]pyrimidine 97%

Supplier: Ambeed
Danger

A1467290-1G A1467290-5G A1467290-25G A1467290-250MG
76662-546EA 74.27 USD
76662-546 77549-798 77567-004 77592-746
2,4,7-Trichloro-8-fluoropyrido[4,3-d]pyrimidine 97%
2,4,7-Trichloro-8-fluoropyrido[4,3-d]pyrimidine

Precaución: For research use only.

Formula: C₇HCl₃FN₃
MW: 252.4603 g/mol
MDL Number: MFCD34167524
CAS Number: 2454396-80-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR