(2R,4R)-Pentane-2,4-diylbis(bis(4-(tert-butyl)phenyl)phosphine) 97+%

Supplier: Ambeed
Warning

A1467218-100MG A1467218-1G A1467218-250MG
76662-540EA 82.76 USD
76662-540 76662-542 76662-544
(2R,4R)-Pentane-2,4-diylbis(bis(4-(tert-butyl)phenyl)phosphine) 97+%
(2R,4R)-Pentane-2,4-diylbis(bis(4-(tert-butyl)phenyl)phosphine)

Precaución: for Research Use Only

Formula: C₄₅H₆₂P₂
MW: 664.9213 g/mol
CAS Number: 1459709-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR