(2S,4S)-Pentane-2,4-diylbis(bis(4-(tert-butyl)phenyl)phosphine) 97+%

Supplier: Ambeed
Warning

A1467216-100MG A1467216-1G A1467216-250MG
76662-534EA 140.04 USD
76662-534 76662-536 76662-538
(2S,4S)-Pentane-2,4-diylbis(bis(4-(tert-butyl)phenyl)phosphine) 97+%
(2S,4S)-Pentane-2,4-diylbis(bis(4-(tert-butyl)phenyl)phosphine)

Precaución: for Research Use Only

Formula: C₄₅H₆₂P₂
MW: 664.9213 g/mol
CAS Number: 911415-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR