Methyl 4-(benzyloxy)-6-(hydroxymethyl)picolinate 95+%

Supplier: Ambeed
Warning

A1467170-100MG A1467170-1G A1467170-250MG A1467170-50MG
76662-526EA 76 USD
76662-526 76662-528 76662-530 76662-532
Methyl 4-(benzyloxy)-6-(hydroxymethyl)picolinate 95+%
Methyl 4-(benzyloxy)-6-(hydroxymethyl)picolinate

Precaución: for Research Use Only

Formula: C₁₅H₁₅NO₄
MW: 273.2839 g/mol
CAS Number: 2098124-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR