(R)-3-(Adamantan-1-yl)-1-(2-hydroxy-1-phenylethyl)-4,5-dihydro-1H-imidazol-3-ium hexafluorophosphate(V) 97%

Supplier: Ambeed
Warning

A1467128-100MG A1467128-50MG
76662-514EA 93 USD
76662-514 76662-516
(R)-3-(Adamantan-1-yl)-1-(2-hydroxy-1-phenylethyl)-4,5-dihydro-1H-imidazol-3-ium hexafluorophosphate(V) 97%
(R)-3-(Adamantan-1-yl)-1-(2-hydroxy-1-phenylethyl)-4,5-dihydro-1H-imidazol-3-ium hexafluorophosphate(V)

Precaución: for Research Use Only

Formula: C₂₁H₂₉F₆N₂OP
CAS Number: 2281885-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR