(E)-3-(4-Bromophenyl)prop-2-en-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

A1467010-100MG A1467010-1G A1467010-250MG
76662-478EA 136.86 USD
76662-478 76662-480 76662-482
(E)-3-(4-Bromophenyl)prop-2-en-1-amine hydrochloride 95%
(E)-3-(4-Bromophenyl)prop-2-en-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁BrClN
MW: 248.5473 g/mol
CAS Number: 2828433-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR