(Z)-((2-Hydroxyethyl)azanediyl)bis(ethane-2,1-diyl) dioleate 98%

Supplier: Ambeed
Warning

A1466984-100G A1466984-25G
76662-474EA 16.98 USD
76662-474 76662-476
(Z)-((2-Hydroxyethyl)azanediyl)bis(ethane-2,1-diyl) dioleate 98%
(Z)-((2-Hydroxyethyl)azanediyl)bis(ethane-2,1-diyl) dioleate

Precaución: for Research Use Only

Formula: C₄₂H₇₉NO₅
MW: 678.0804 g/mol
MDL Number: MFCD23160511
CAS Number: 54999-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR