2-(2-Bromo-3-methylphenyl)acetamide 95%

Supplier: Ambeed
Warning

A1466870-100MG A1466870-250MG A1466870-5G
76662-456EA 41 USD
76662-456 76662-458 76662-460
2-(2-Bromo-3-methylphenyl)acetamide 95%
2-(2-Bromo-3-methylphenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
MDL Number: MFCD28132211
CAS Number: 1824058-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR