4-Mercaptobenzoic acid 90+% (by HPLC)
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Thiobenzoic acid
A1466091-1G
A1466091-25G
A1466091-5G
76662-450EA
57.29
USD
76662-450
76662-452
76662-454
4-Mercaptobenzoic acid 90+% (by HPLC)
4-Mercaptobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₆O₂S MW: 154.1863 g/mol |
MDL Number:
MFCD00016617 CAS Number: 1074-36-8 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...