trans-Bis(tricyclohexylphosphine)(2-methylphenyl)nickel(II) chloride 98%

Supplier: Ambeed
Danger

A1465845-100MG A1465845-1G
76662-448EA 115 USD
76662-448 77875-032
trans-Bis(tricyclohexylphosphine)(2-methylphenyl)nickel(II) chloride 98%
trans-Bis(tricyclohexylphosphine)(2-methylphenyl)nickel(II) chloride

Precaución: for Research Use Only

Formula: C₄₃H₇₅ClNiP₂
MW: 748.1495 g/mol
MDL Number: MFCD31619189
CAS Number: 123839-28-1
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR