2-Chloro-N-(6-methoxyquinolin-4-yl)-5-nitrobenzamide 98%

Supplier: Ambeed
Warning

Synonyms: SR 16832

A1465496-25MG A1465496-100MG A1465496-250MG A1465496-50MG
76662-438EA 80 USD
76662-438 77875-017 77875-018 77875-019
2-Chloro-N-(6-methoxyquinolin-4-yl)-5-nitrobenzamide 98%
2-Chloro-N-(6-methoxyquinolin-4-yl)-5-nitrobenzamide

Precaución: for Research Use Only

Formula: C₁₇H₁₂ClN₃O₄
MDL Number: MFCD31656711
CAS Number: 2088135-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR