2-(2-Aminothiazol-4-yl)-N-(3-chloro-4-methylphenyl)acetamide 95%

Supplier: Ambeed
Warning

A1465152-100MG A1465152-1G A1465152-250MG
76662-432EA 41.38 USD
76662-432 76662-434 76662-436
2-(2-Aminothiazol-4-yl)-N-(3-chloro-4-methylphenyl)acetamide 95%
2-(2-Aminothiazol-4-yl)-N-(3-chloro-4-methylphenyl)acetamide

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClN₃OS
MW: 281.7612 g/mol
MDL Number: MFCD17127074
CAS Number: 1327839-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR