2-(Methoxycarbonyl)cyclohexanecarboxylic acid 98%

Supplier: Ambeed
Warning

A1465056-1G A1465056-25G A1465056-5G
76662-426EA 90.18 USD
76662-426 76662-428 76662-430
2-(Methoxycarbonyl)cyclohexanecarboxylic acid 98%
2-(Methoxycarbonyl)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₄O₄
MDL Number: MFCD00154581
CAS Number: 7719-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR