tert-Butyl (4-bromo-7-fluorobenzo[d]thiazol-2-yl)carbamate 95%

Supplier: Ambeed
Warning

A1464884-100MG A1464884-1G A1464884-250MG A1464884-25MG
76662-410EA 235 USD
76662-410 76662-412 76662-414 76662-416
tert-Butyl (4-bromo-7-fluorobenzo[d]thiazol-2-yl)carbamate 95%
tert-Butyl (4-bromo-7-fluorobenzo[d]thiazol-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₂BrFN₂O₂S
MDL Number: MFCD34186958
CAS Number: 2649788-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR