(4R,4'R,5S,5'S)-2,2'-(1,3-Bis(4-(adamantan-1-yl)phenyl)propane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) 97%
Supplier: Ambeed
|
Warning
|
A1464846-100MG
A1464846-250MG
76662-394EA
192.03
USD
76662-394
77624-232
(4R,4'R,5S,5'S)-2,2'-(1,3-Bis(4-(adamantan-1-yl)phenyl)propane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) 97%
(4R,4'R,5S,5'S)-2,2'-(1,3-Bis(4-(adamantan-1-yl)phenyl)propane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole)
Precaución: for Research Use Only
Formula:
C₆₅H₆₆N₂O₂ MW: 907.23 g/mol |
CAS Number:
2647945-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...