(4S,4'S,5R,5'R)-2,2'-(1,3-Bis(4-(adamantan-1-yl)phenyl)propane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) 97%

Supplier: Ambeed
Warning

A1464844-100MG A1464844-250MG
76662-392EA 202.64 USD
76662-392 77597-354
(4S,4'S,5R,5'R)-2,2'-(1,3-Bis(4-(adamantan-1-yl)phenyl)propane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole) 97%
(4S,4'S,5R,5'R)-2,2'-(1,3-Bis(4-(adamantan-1-yl)phenyl)propane-2,2-diyl)bis(4,5-diphenyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₆₅H₆₆N₂O₂
MW: 907.2318 g/mol
CAS Number: 2582969-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR