Ammonium trifluoromethanesulfonate 98+%

Supplier: Ambeed
Warning

A1464688-25G A1464688-1G A1464688-5G
76662-372EA 109 USD
76662-372 77597-344 77597-346
Ammonium trifluoromethanesulfonate 98+%
Ammonium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: CH₄F₃NO₃S
MDL Number: MFCD00075332
CAS Number: 38542-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR