2,5-Difluorophenol 99%

Supplier: Ambeed
Danger

A1464682-1G A1464682-25G A1464682-5G
76662-366EA 6.37 USD
76662-366 76662-368 76662-370
2,5-Difluorophenol 99%
2,5-Difluorophenol

Precaución: for Research Use Only

Formula: C₆H₄F₂O
MW: 130.09 g/mol
MDL Number: MFCD00042501
CAS Number: 2713-31-7
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR