Tetrabutylammonium sulfate 50% (w/w) in water
Supplier: Ambeed
|
Warning
|
A1464572-5G
A1464572-25G
76662-350EA
10
USD
76662-350
77624-230
Tetrabutylammonium sulfate 50% (w/w) in water
Tetrabutylammonium sulphate
Precaución: for Research Use Only
Formula:
C₃₂H₇₂N₂O₄S |
MDL Number:
MFCD00216632 CAS Number: 2472-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...