BAPTA tetraethyl ester 97+%

Supplier: Ambeed
Warning

Synonyms: Tetraethyl 2,2',2'',2'''-(((ethane-1,2-diylbis(oxy))bis(2,1-phenylene))bis(azanetriyl))tetraacetate

A1464540-1G A1464540-25G A1464540-5G
76662-330EA 21.22 USD
76662-330 76662-332 76662-334
BAPTA tetraethyl ester 97+%
BAPTA tetraethyl ester

Precaución: for Research Use Only

Formula: C₃₀H₄₀N₂O₁₀
MW: 588.66 g/mol
MDL Number: MFCD00532663
CAS Number: 73630-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR