Creatine phosphate disodium salt 98+%

Supplier: Ambeed
Warning

Synonyms: Sodium creatine phosphate dibasic, N-Methyl-N-(N-phosphonocarbamimidoyl)glycine, disodium salt, Sodium creatine phosphate, Phosphocreatine disodium salt

A1464538-100G A1464538-25G A1464538-5G
76662-324EA 173 USD
76662-324 76662-326 76662-328
Creatine phosphate disodium salt 98+%
Creatine phosphate disodium salt

Precaución: for Research Use Only

Formula: C₄H₈N₃O₅P·2Na
MW: 255.077 g/mol
MDL Number: MFCD00014351
CAS Number: 922-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR