3,3'-Dimethylnaphthidine 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Diamino-3,3'dimethyl-1,1'-binaphthyl, 3,3'-Dimethyl-[1,1'-binaphthalene]-4,4'-diamine, 4-(4-amino-3-methylnaphthalen-1-yl)-2-methylnaphthalen-1-amine

A1464424-1G A1464424-250MG A1464424-25G A1464424-5G
76662-308EA 71.09 USD
76662-308 76662-310 76662-312 76662-314
3,3'-Dimethylnaphthidine 97%
3,3'-Dimethylnaphthidine

Precaución: for Research Use Only

Formula: C₂₂H₂₀N₂
MW: 312.42 g/mol
MDL Number: MFCD00042706
CAS Number: 13138-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR