3-(3-Pyridyl)-L-alanine ≥99%, ee 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-Amino-3-(pyridin-3-yl)propanoic acid, H-β-(3-Pyridyl)-Ala-OH, H-3-Pal-OH
A1464269-1G
A1464269-250MG
A1464269-25G
A1464269-5G
76662-300EA
65.78
USD
76662-300
76662-302
76662-304
76662-306
3-(3-Pyridyl)-L-alanine ≥99%, ee 98%
3-(3-Pyridyl)-L-alanine
Precaución: for Research Use Only
Formula:
C₈H₁₀N₂O₂ MW: 166.18 g/mol |
MDL Number:
MFCD00079676 CAS Number: 64090-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...