(11bS)-4-Hydroxy-2,6-bis(4-nitrophenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 97%
Supplier: Ambeed
|
Warning
|
A1464199-100MG
A1464199-1G
A1464199-250MG
76662-294EA
26
USD
76662-294
76662-296
76662-298
(11bS)-4-Hydroxy-2,6-bis(4-nitrophenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 97%
(11bS)-4-Hydroxy-2,6-bis(4-nitrophenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide
Precaución: for Research Use Only
Formula:
C₃₂H₂₇N₂O₈P |
CAS Number:
2129645-31-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...