Diethyl-5-aminoisophthalate 97%

Supplier: Ambeed
Warning

A1464181-100MG A1464181-1G A1464181-250MG A1464181-25G A1464181-5G
76662-284EA 22 USD
76662-284 76662-286 76662-288 76662-290 76662-292
Diethyl-5-aminoisophthalate 97%
Diethyl-5-aminoisophthalate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₄
MDL Number: MFCD00127691
CAS Number: 42122-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR