Bis(tricyclohexylphosphine)palladium(0) 98%
Supplier: Ambeed
|
Warning
|
A1464082-1G
A1464082-250MG
A1464082-100MG
A1464082-5G
76662-270EA
89
USD
76662-270
76662-272
77622-202
77875-001
Bis(tricyclohexylphosphine)palladium(0) 98%
Bis(tricyclohexylphosphine)palladium(0)
Precaución: for Research Use Only
Formula:
C₃₆H₆₆P₂Pd |
MDL Number:
MFCD01073796 CAS Number: 33309-88-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...