Calcium acetate monohydrate ≥98%
Supplier: Ambeed
|
Warning
|
A1464042-100G
A1464042-25G
A1464042-500G
76662-264EA
6
USD
76662-264
76662-266
76662-268
Calcium acetate monohydrate ≥98%
Calcium acetate monohydrate
Precaución: for Research Use Only
Formula:
(H₃CCOO)₂Ca·1H₂O |
MDL Number:
MFCD00150019 CAS Number: 5743-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...