tert-butyl (1-Imino-1-oxidotetrahydro-2H-thiopyran-4-yl)(methyl)carbamate 97%

Supplier: Ambeed
Warning

A1464032-1G A1464032-100MG A1464032-250MG
76662-262EA 101 USD
76662-262 77874-999 77875-000
tert-butyl (1-Imino-1-oxidotetrahydro-2H-thiopyran-4-yl)(methyl)carbamate 97%
tert-butyl (1-Imino-1-oxidotetrahydro-2H-thiopyran-4-yl)(methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₃S
MDL Number: MFCD34563714
CAS Number: 2816674-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR