Adenosine-5'-diphosphate monopotassium salt 97%

Supplier: Ambeed
Danger

Synonyms: ADP.K, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl trihydrogen diphosphate monopotassium salt, 5'-ADP-K

A1464002-1G A1464002-250MG A1464002-25G A1464002-5G
76662-254EA 15.92 USD
76662-254 76662-256 76662-258 76662-260
Adenosine-5'-diphosphate monopotassium salt 97%
Adenosine-5'-diphosphate monopotassium salt

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₅O₁₀P₂K
MW: 465.29 g/mol
MDL Number: MFCD00066472
CAS Number: 70285-70-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR