3-(4-Fluorophenyl)-1-methyl-1H-pyrazol-4-amine hydrochloride 97%

Supplier: Ambeed
Warning

A1463902-100MG A1463902-250MG
76662-250EA 105 USD
76662-250 76662-252
3-(4-Fluorophenyl)-1-methyl-1H-pyrazol-4-amine hydrochloride 97%
3-(4-Fluorophenyl)-1-methyl-1H-pyrazol-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClFN₃
MDL Number: MFCD34186955
CAS Number: 2703756-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR