Sodium tungstate(VI) 97%

Supplier: Ambeed
Warning

Synonyms: NaDT

A1463870-100MG A1463870-1G A1463870-50MG A1463870-5G A1463870-250MG
76662-242EA 75.33 USD
76662-242 76662-244 76662-246 76662-248 77597-328
Sodium tungstate(VI) 97%
Sodium tungstate(VI)

Precaución: for Research Use Only

Formula: Na₄O₃₂W₁₀
MW: 2442.34 g/mol
CAS Number: 126752-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR