(Oxybis(2,1-phenylene))bis(bis(4-methoxy-3,5-dimethylphenyl)phosphine) 97%

Supplier: Ambeed
Warning

A1463828-100MG A1463828-250MG A1463828-50MG A1463828-1G
76662-232EA 255 USD
76662-232 76662-234 76662-236 77874-998
(Oxybis(2,1-phenylene))bis(bis(4-methoxy-3,5-dimethylphenyl)phosphine) 97%
(Oxybis(2,1-phenylene))bis(bis(4-methoxy-3,5-dimethylphenyl)phosphine)

Precaución: for Research Use Only

Formula: C₄₈H₅₂O₅P₂
CAS Number: 2054939-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR