(S)-1-(1-Hydroxy-3,3-dimethylbutan-2-yl)-3-mesityl-4,5-dihydro-1H-imidazol-3-ium hexafluorophosphate(V) 97%

Supplier: Ambeed
Warning

A1463782-100MG A1463782-1G A1463782-250MG
76662-220EA 78.51 USD
76662-220 77622-198 77622-200
(S)-1-(1-Hydroxy-3,3-dimethylbutan-2-yl)-3-mesityl-4,5-dihydro-1H-imidazol-3-ium hexafluorophosphate(V) 97%
(S)-1-(1-Hydroxy-3,3-dimethylbutan-2-yl)-3-mesityl-4,5-dihydro-1H-imidazol-3-ium hexafluorophosphate(V)

Precaución: for Research Use Only

Formula: C₁₈H₂₉F₆N₂OP
MW: 434.4 g/mol
CAS Number: 850469-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR