2-Amino-5-bromo-3-methylbenzonitrile 97%

Supplier: Ambeed
Warning

A1463768-100MG A1463768-1G A1463768-250MG
76662-214EA 68.96 USD
76662-214 76662-216 76662-218
2-Amino-5-bromo-3-methylbenzonitrile 97%
2-Amino-5-bromo-3-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.0586 g/mol
MDL Number: MFCD22024490
CAS Number: 1093966-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR