(5,6-Dimethylpyridin-2-yl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A1463644-100MG A1463644-1G A1463644-250MG A1463644-5G
76662-196EA 61 USD
76662-196 76662-198 76662-200 76662-202
(5,6-Dimethylpyridin-2-yl)methanamine hydrochloride 97%
(5,6-Dimethylpyridin-2-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₃ClN₂
MDL Number: MFCD20923685
CAS Number: 2448337-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR