5-Bromo-2-chloro-1,3-diiodobenzene 95%

Supplier: Ambeed
Warning

A1463624-100MG A1463624-1G A1463624-250MG A1463624-5G
76662-188EA 20.16 USD
76662-188 76662-190 76662-192 76662-194
5-Bromo-2-chloro-1,3-diiodobenzene 95%
5-Bromo-2-chloro-1,3-diiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClI₂
MW: 443.2460 g/mol
CAS Number: 2383588-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR