3-(2-Aminoethyl)-1H-indole-6-carbonitrile hydrochloride 97%

Supplier: Ambeed
Warning

A1462878-100MG A1462878-1G A1462878-250MG
76662-132EA 79.57 USD
76662-132 76662-134 76662-136
3-(2-Aminoethyl)-1H-indole-6-carbonitrile hydrochloride 97%
3-(2-Aminoethyl)-1H-indole-6-carbonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClN₃
MW: 221.6861 g/mol
MDL Number: MFCD34178764
CAS Number: 2703752-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR