3-(2-Chloropropanoyl)spiro[benzo[e][1,3]oxazine-2,1'-cyclohexan]-4(3H)-one 97%

Supplier: Ambeed
Warning

A1462822-100G A1462822-25G A1462822-500G
76662-126EA 31.83 USD
76662-126 76662-128 76662-130
3-(2-Chloropropanoyl)spiro[benzo[e][1,3]oxazine-2,1'-cyclohexan]-4(3H)-one 97%
3-(2-Chloropropanoyl)spiro[benzo[e][1,3]oxazine-2,1'-cyclohexan]-4(3H)-one

Precaución: for Research Use Only

Formula: C₁₆H₁₈ClNO₃
MW: 307.7720 g/mol
MDL Number: MFCD32201942
CAS Number: 1332594-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR