N,N-Dimethyl-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-2-amine dihydrochloride 97%

Supplier: Ambeed
Warning

A1462788-100MG A1462788-1G A1462788-250MG
76662-112EA 41.38 USD
76662-112 76662-114 76662-116
N,N-Dimethyl-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-2-amine dihydrochloride 97%
N,N-Dimethyl-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-2-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₆Cl₂N₄
MW: 251.1561 g/mol
MDL Number: MFCD28892716
CAS Number: 2098031-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR