L-Lysine ethyl ester diisocyanate 97%

Supplier: Ambeed
Danger

A1462758-1G A1462758-5G
76662-108EA 13 USD
76662-108 76662-110
L-Lysine ethyl ester diisocyanate 97%
L-Lysine ethyl ester diisocyanate

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₄
MDL Number: MFCD08276480
CAS Number: 45172-15-4
UN: 2206
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR