Bicyclo[1.1.1]pentane-1-sulfonamide 97%

Supplier: Ambeed
Warning

A1462746-100MG A1462746-250MG A1462746-50MG
76662-102EA 578.2 USD
76662-102 76662-104 76662-106
Bicyclo[1.1.1]pentane-1-sulfonamide 97%
Bicyclo[1.1.1]pentane-1-sulfonamide

Precaución: for Research Use Only

Formula: C₅H₉NO₂S
MW: 147.1955 g/mol
MDL Number: MFCD32678744
CAS Number: 2413074-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR